Please login to your online account to display your discounted pricing

3-Chloropropiophenone, 90%, tech., ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC369980100

 View more versions of this product

Catalog No. AC369980100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥88%
Purity Grade Technical Grade
Physical Form Crystalline Solid
Formula Weight 168.62
Quantity 10g
Color Beige to brown
Beilstein 07, 302
CAS Min % 88.0
CAS Max % 100.0
Linear Formula C6H5COCH2CH2Cl
Name Note tech., 90%
Assay Percent Range 90%
Molecular Formula C9H9ClO
MDL Number MFCD00000990
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material 3-Chloropropiophenone
Packaging Glass bottle
CAS 936-59-4
Synonym 3-Chloro-1-phenyl-1-propanone
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Chloropropiophenone
Name Note tech., 90%
CAS 936-59-4
CAS Min % 88.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C9H9ClO
Linear Formula C6H5COCH2CH2Cl
Formula Weight 168.62
MDL Number MFCD00000990
Synonym 3-Chloro-1-phenyl-1-propanone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents