Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloropropionyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Chloropropionyl chloride
CAS 625-36-5
Molecular Formula C3H4Cl2O
Linear Formula ClCH2CH2COCl
Formula Weight 126.97
MDL Number MFCD00000747
UN Number UN3389
Beilstein 635814
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2569822

  SDS
alfa aesar™
B2569822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.97
MDL Number MFCD00000747
Assay 97%
RTECS Number UC3934000
Linear Formula ClCH2CH2COCl
Beilstein 635814
CAS 625-36-5
Chemical Name or Material 3-Chloropropionyl chloride
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Quantity 100g
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.
Melting Point −32°C
Boiling Point 143° to 145°C
Health Hazard 2 GHS H Statement
H301-H330-H314-H318-H226
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Flash Point 59°C (138°F)
Molecular Formula C3H4Cl2O
UN Number UN3389

Applications
3-Chlorpropionyl chloride can be used in the production process of pharma ingredients.

Solubility
Reacts violently with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases, active metals.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H330-H314-H318-H226
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.