missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloropropionyl chloride, 97%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals B2569822

Catalog No. AAB2569822



Description

Description

3-Chlorpropionyl chloride can be used in the production process of pharma ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorpropionyl chloride can be used in the production process of pharma ingredients.

Solubility
Reacts violently with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ mois
Specifications

Specifications

3-Chloropropionyl chloride
-32°C
143°C to 145°C
C3H4Cl2O
ClCH2CH2COCl
100 g
635814
3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
INUNLMUAPJVRME-UHFFFAOYSA-N
3-chloropropanoyl chloride
69364
97%
625-36-5
1.325
59°C (138°F)
1.457
MFCD00000747
UN3389
Moisture sensitive
Reacts violently with water.
ClCCC(Cl)=O
126.96
126.97

Chemical Identifiers

625-36-5
126.96
INUNLMUAPJVRME-UHFFFAOYSA-N
69364
ClCCC(Cl)=O
C3H4Cl2O
MFCD00000747
3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
3-chloropropanoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H314-H318-H226
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c

H226-H290-H302-H314-H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 210-890-4

RTECSNumber : UC3934000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only