missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloropropionic Acid 98%, ACROS Organics™

Manufacturer:  Acros Organics 158155000

 View more versions of this product

Catalog No. AC158155000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Chloropropionic acid
203.0°C to 205.0°C
37.0°C to 41.0°C
98%
97.5
Authentic
98%
C3H5ClO2
MFCD00002764
15, 2161
C(CCl)C(=O)O
108.521
108.52
125°C
White through Beige
500g
107-94-8
100.0
Glass bottle
98%
ClCH2CH2CO2H
02, 249
QEYMMOKECZBKAC-UHFFFAOYSA-N
3-chloropropanoic acid
7899
Crystalline Low Melting Mass

Chemical Identifiers

107-94-8
108.521
QEYMMOKECZBKAC-UHFFFAOYSA-N
C3H5ClO2
MFCD00002764
7899
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Corrosive to the respiratory tract.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-534-4

RTECSNumber : UE8750000

TSCA : TSCA

SDS Documents