missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloropropionic acid, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 158151000

 View more versions of this product

Catalog No. AC158151000



Specifications

Specifications

3-Chloropropionic acid
Authentic
203.0°C to 205.0°C
37.0°C to 41.0°C
98%
97.5
98%
ClCH2CH2CO2H
02, 249
.beta.-chloropropionic acid
QEYMMOKECZBKAC-UHFFFAOYSA-N
3-chloropropanoic acid
7899
Crystalline Low Melting Mass
125°C
Glass bottle
White to Beige
100g
107-94-8
100.0
C3H5ClO2
MFCD00002764
15, 2161
Solubility in water: freely soluble. Other solubilities: soluble in alcohol,chloroform and ether,miscible with ether
C(CCl)C(=O)O
108.52
108.52
98%

Chemical Identifiers

107-94-8
108.52
QEYMMOKECZBKAC-UHFFFAOYSA-N
7899
C(CCl)C(=O)O
C3H5ClO2
MFCD00002764
.beta.-chloropropionic acid
3-chloropropanoic acid
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Corrosive to the respiratory tract.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-534-4

RTECSNumber : UE8750000

TSCA : TSCA

SDS Documents