Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloropropionic acid, 98%

$35.55 - $35.55

Chemical Identifiers

Chemical Name or Material 3-Chloropropionic acid
CAS 107-94-8
Molecular Formula ClCH2CH2CO2H
Formula Weight 108.53
MDL Number MFCD00002764
UN Number UN3261
Beilstein 1098495
Merck Index 14,2159
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2174422

  SDS
alfa aesar™
B2174422
100g Each for $35.55

AAB2174436

  SDS
alfa aesar™
B2174436
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.53
Chemical Name or Material 3-Chloropropionic acid
TSCA TSCA
Assay 98%
Molecular Formula ClCH2CH2CO2H
MDL Number MFCD00002764
CAS 107-94-8
UN Number UN3261
Beilstein 1098495
Merck Index 14,2159
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number UE8750000
Flash Point 122°C (251°F)
Boiling Point 203° to 205°C
Melting Point 37° to 41°C
Quantity 100g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.