missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloropropionaldehyde Diethyl Acetal (stabilized with CaCO3), TCI America™

$79.45 - $233.75

Chemical Identifiers

CAS 35573-93-4
Molecular Formula C7H15ClO2
Molecular Weight (g/mol) 166.645
MDL Number MFCD00000994
InChI Key NXHONHDWVLPPCS-UHFFFAOYSA-N
Synonym 3-chloropropionaldehyde diethyl acetal, propane, 3-chloro-1,1-diethoxy, 3-chloropropionaldehyde diethylacetal, 1-chloro-3,3-diethoxypropane, .beta.-chloropropionaldehyde diethyl acetal, 3,3-diethoxypropylchloride, 3,3-diethoxypropyl chloride, 3-ethoxy-4-oxahexyl chloride, 3-chloro-1,1-diethoxy-propane, 3-chloropropanal diethyl acetal
PubChem CID 96217
IUPAC Name 3-chloro-1,1-diethoxypropane
SMILES CCOC(CCCl)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C126825G View Documents TCI America
C126825G
25g Each for $233.75
 
C12685G View Documents TCI America
C12685G
5g Each for $79.45
 
Specifications

Chemical Identifiers

35573-93-4
166.645
NXHONHDWVLPPCS-UHFFFAOYSA-N
96217
CCOC(CCCl)OCC
C7H15ClO2
MFCD00000994
3-chloropropionaldehyde diethyl acetal, propane, 3-chloro-1,1-diethoxy, 3-chloropropionaldehyde diethylacetal, 1-chloro-3,3-diethoxypropane, .beta.-chloropropionaldehyde diethyl acetal, 3,3-diethoxypropylchloride, 3,3-diethoxypropyl chloride, 3-ethoxy-4-oxahexyl chloride, 3-chloro-1,1-diethoxy-propane, 3-chloropropanal diethyl acetal
3-chloro-1,1-diethoxypropane

Specifications

74°C
25g
C7H15ClO2
1993
NXHONHDWVLPPCS-UHFFFAOYSA-N
3-chloro-1,1-diethoxypropane
96217
Clear Liquid at 20°C
3-Chloropropionaldehyde Diethyl Acetal (stabilized with CaCO3)
Yellow
35573-93-4
MFCD00000994
3-chloropropionaldehyde diethyl acetal, propane, 3-chloro-1,1-diethoxy, 3-chloropropionaldehyde diethylacetal, 1-chloro-3,3-diethoxypropane, .beta.-chloropropionaldehyde diethyl acetal, 3,3-diethoxypropylchloride, 3,3-diethoxypropyl chloride, 3-ethoxy-4-oxahexyl chloride, 3-chloro-1,1-diethoxy-propane, 3-chloropropanal diethyl acetal
CCOC(CCCl)OCC
166.645
166.65
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator