Learn More
3-Chloropropanesulfonyl chloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1063118
Description
3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.
Solubility
Soluble in dichloromethane, ethyl Acetate and hexane.
Notes
Store at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Keep away from alcohols, strong bases, oxidizing agents.
Specifications
3-Chloropropanesulfonyl chloride | |
1.456 | |
>110°C (230°F) | |
1.49 | |
MFCD00007463 | |
UN3265 | |
Moisture sensitive | |
Soluble in dichloromethane,ethyl Acetate and hexane. | |
C(CS(=O)(=O)Cl)CCl | |
177.039 | |
177.05 |
1633-82-5 | |
70°C (0.5 mmHg) | |
C3H6Cl2O2S | |
Cl(CH2)3SO2Cl | |
50 g | |
1754119 | |
3-chloropropanesulfonyl chloride, 3-chloro-1-propanesulfonyl chloride, 3-chloropropanesulfonylchloride, 3-chloropropylsulfonyl chloride, 3-chloropropanesulphonyl chloride, 1-propanesulfonyl chloride, 3-chloro, propanesulfonyl chloride, chloro, 3-chloro-propane-1-sulfonyl chloride, gamma-chloropropanesulfonyl chloride, 3-chloropropylsulfonylchloride | |
GPKDGVXBXQTHRY-UHFFFAOYSA-N | |
3-chloropropane-1-sulfonyl chloride | |
15410 | |
≥98% |
Chemical Identifiers
1633-82-5 | |
177.039 | |
GPKDGVXBXQTHRY-UHFFFAOYSA-N | |
15410 | |
C(CS(=O)(=O)Cl)CCl |
C3H6Cl2O2S | |
MFCD00007463 | |
3-chloropropanesulfonyl chloride, 3-chloro-1-propanesulfonyl chloride, 3-chloropropanesulfonylchloride, 3-chloropropylsulfonyl chloride, 3-chloropropanesulphonyl chloride, 1-propanesulfonyl chloride, 3-chloro, propanesulfonyl chloride, chloro, 3-chloro-propane-1-sulfonyl chloride, gamma-chloropropanesulfonyl chloride, 3-chloropropylsulfonylchloride | |
3-chloropropane-1-sulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 216-646-3
RTECSNumber : TZ6670000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only