missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorophthalic Anhydride 98.0+%, TCI America™

Supplier:  TCI America C348925G

 View more versions of this product

Catalog No. C348925G



Description

Description

Specifications

Specifications

3-Chlorophthalic Anhydride
128°C
170°C
MFCD00023107
3-chlorophthalic anhydride, 4-chloroisobenzofuran-1,3-dione, 3-chlorophthalic acid anhydride, unii-vo7nxt0137, 1,3-isobenzofurandione, chloro, 4-chloro-1,3-dihydro-2-benzofuran-1,3-dione, 4-chloranyl-2-benzofuran-1,3-dione, 3clpa, pubchem19464, 3-chlorophthalicanhydride
ClC1=CC=CC2=C1C(=O)OC2=O
182.56
182.56
Crystalline Powder
117-21-5
White
C8H3ClO3
25 g
UERPUZBSSSAZJE-UHFFFAOYSA-N
4-chloro-1,3-dihydro-2-benzofuran-1,3-dione
67014
≥98.0% (GC)

Chemical Identifiers

117-21-5
182.56
UERPUZBSSSAZJE-UHFFFAOYSA-N
67014
ClC1=CC=CC2=C1C(=O)OC2=O
C8H3ClO3
MFCD00023107
3-chlorophthalic anhydride, 4-chloroisobenzofuran-1,3-dione, 3-chlorophthalic acid anhydride, unii-vo7nxt0137, 1,3-isobenzofurandione, chloro, 4-chloro-1,3-dihydro-2-benzofuran-1,3-dione, 4-chloranyl-2-benzofuran-1,3-dione, 3clpa, pubchem19464, 3-chlorophthalicanhydride
4-chloro-1,3-dihydro-2-benzofuran-1,3-dione
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0062

TSCA : No

SDS
Documents

Documents

Promotions

Promotions