Please login to your online account to display your discounted pricing

3-Chlorophthalic Anhydride, +95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Chlorophthalic anhydride
CAS 117-21-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0% (morpholine coupling)
Assay Percent Range 95+%
Molecular Formula C8H3ClO3
Formula Weight 182.56
MDL Number MFCD00023107
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381290250

  SDS
thermo fisher scientific
AC381290250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC381290010

  SDS
fisher scientific acros
AC381290010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC381290050

  SDS
fisher scientific acros
AC381290050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95.0% (morpholine coupling)
Physical Form Crystalline Powder and Chunks
Formula Weight 182.56
Quantity 25g
Color White or pale yellow
Melting Point 123° to 126°C
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 3-Chlorophthalic anhydride
Assay Percent Range 95+%
MDL Number MFCD00023107
CAS 117-21-5
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C8H3ClO3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.