Please login to your online account to display your discounted pricing

3-Chlorophthalic Anhydride, +95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC381290010

 View more versions of this product

Catalog No. AC381290010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95.0% (morpholine coupling)
Physical Form Crystalline Powder and Chunks
Formula Weight 182.56
Quantity 1g
Color White or pale yellow
Melting Point 123° to 126°C
CAS Max % 100.0
CAS Min % 95.0
Infrared Spectrum Authentic
Chemical Name or Material 3-Chlorophthalic anhydride
Molecular Formula C8H3ClO3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
CAS 117-21-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00023107
Assay Percent Range 95+%

Specifications

Chemical Name or Material 3-Chlorophthalic anhydride
CAS 117-21-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0% (morpholine coupling)
Assay Percent Range 95+%
Molecular Formula C8H3ClO3
Formula Weight 182.56
MDL Number MFCD00023107
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents