Learn More
3-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H51161AE
Description
3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1'-DiBoc-2,2'-Biaziridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1′-DiBoc-2,2′-Biaziridine.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents. Reacts with water.
Specifications
3-Chlorophenylmagnesium bromide | |
100 mL | |
MFCD00672003 | |
3-chlorophenylmagnesium bromide, 3-chlorophenyl magnesium bromide, magnesium,bromo 3-chlorophenyl, m-chlorophenylmagnesium bromide, m-chlorophenyl magnesium bromide, 3-chloro phenyl magnesium bromide, 3-chloro-phenyl magnesium bromide, ziyruwptupfrqu-uhfffaoysa-m, grignard reagent | |
ClC1=CC([Mg]Br)=CC=C1 | |
215.76 | |
215.76 |
36229-42-2 | |
C6H4BrClMg | |
UN2924 | |
ZIYRUWPTUPFRQU-UHFFFAOYSA-M | |
magnesium;chlorobenzene;bromide | |
12252967 | |
1M in MeTHF |
Chemical Identifiers
36229-42-2 | |
215.76 | |
ZIYRUWPTUPFRQU-UHFFFAOYSA-M | |
12252967 | |
ClC1=CC([Mg]Br)=CC=C1 |
C6H4BrClMg | |
MFCD00672003 | |
3-chlorophenylmagnesium bromide, 3-chlorophenyl magnesium bromide, magnesium,bromo 3-chlorophenyl, m-chlorophenylmagnesium bromide, m-chlorophenyl magnesium bromide, 3-chloro phenyl magnesium bromide, 3-chloro-phenyl magnesium bromide, ziyruwptupfrqu-uhfffaoysa-m, grignard reagent | |
magnesium;chlorobenzene;bromide |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only