missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chlorophenylisothiocyanate, 98%, ACROS Organics™

Chemical Identifiers

CAS 2392-68-9
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.63
MDL Number MFCD00004805
InChI Key WGXCKFMVBAOIFH-UHFFFAOYSA-N
Synonym 1-chloro-3-isothiocyanato-benzene
PubChem CID 75449
IUPAC Name 1-chloro-3-isothiocyanatobenzene
SMILES C1=CC(=CC(=C1)Cl)N=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC292250250 SDS View Documents Thermo Fisher Scientific
AC292250250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

2392-68-9
169.63
WGXCKFMVBAOIFH-UHFFFAOYSA-N
75449
C1=CC(=CC(=C1)Cl)N=C=S
C7H4ClNS
MFCD00004805
1-chloro-3-isothiocyanato-benzene
1-chloro-3-isothiocyanatobenzene

Specifications

1.2920g/mL
Authentic
1.6580 to 1.66
249°C to 250°C
25g
97.5
97.5% min. (GC)
ClC6H4NCS
1-chloro-3-isothiocyanato-benzene
C1=CC(=CC(=C1)Cl)N=C=S
169.63
169.63
98%
>110°C
Glass bottle
1.292
Yellow
2392-68-9
100.0
C7H4ClNS
MFCD00004805
WGXCKFMVBAOIFH-UHFFFAOYSA-N
1-chloro-3-isothiocyanatobenzene
75449
Liquid
3-Chlorophenylisothiocyanate, 98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if inhaled.
Causes skin irritation.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger