missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chlorophenylisothiocyanate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC292250250

 View more versions of this product

Catalog No. AC292250250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Chlorophenylisothiocyanate
>110°C
Glass bottle
1.292
Yellow
98%
97.5
97.5% min. (GC)
ClC6H4NCS
1-chloro-3-isothiocyanato-benzene, 3-chlorobenzenisothiocyanate, 3-chlorophenyl isothiocyanate, 3-chlorophenylisothiocyanate, acmc-1cfb8, benzene, 1-chloro-3-isothiocyanato, isothiocyanic acid, 3-chlorophenyl ester, isothiocyanic acid, m-chlorophenyl ester, m-chlorophenyl isothiocyanate, wln: scnr cg
C1=CC(=CC(=C1)Cl)N=C=S
169.63
169.63
98%
1.2920g/mL
Authentic
1.6580 to 1.66
249°C to 250°C
25g
2392-68-9
100.0
C7H4ClNS
MFCD00004805
WGXCKFMVBAOIFH-UHFFFAOYSA-N
1-chloro-3-isothiocyanatobenzene
75449
Liquid

Chemical Identifiers

2392-68-9
169.63
WGXCKFMVBAOIFH-UHFFFAOYSA-N
75449
C1=CC(=CC(=C1)Cl)N=C=S
C7H4ClNS
MFCD00004805
1-chloro-3-isothiocyanato-benzene, 3-chlorobenzenisothiocyanate, 3-chlorophenyl isothiocyanate, 3-chlorophenylisothiocyanate, acmc-1cfb8, benzene, 1-chloro-3-isothiocyanato, isothiocyanic acid, 3-chlorophenyl ester, isothiocyanic acid, m-chlorophenyl ester, m-chlorophenyl isothiocyanate, wln: scnr cg
1-chloro-3-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if inhaled.
Causes skin irritation.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

SDS Documents