missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorophenylhydrazine Sulfate, TCI America™

$94.72 - $247.50

Chemical Identifiers

CAS 41713-37-5
Molecular Formula C6H9ClN2O4S
Molecular Weight (g/mol) 240.658
MDL Number MFCD00050933
InChI Key ZQMWFTLJZQAIHV-UHFFFAOYSA-N
PubChem CID 20542670
IUPAC Name (3-chlorophenyl)hydrazine;sulfuric acid
SMILES C1=CC(=CC(=C1)Cl)NN.OS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C02595G View Documents TCI America
C02595G
5g Each for $94.72
 
C025925G View Documents TCI America
C025925G
25g Each for $247.50
 
Specifications

Chemical Identifiers

41713-37-5
240.658
ZQMWFTLJZQAIHV-UHFFFAOYSA-N
(3-chlorophenyl)hydrazine;sulfuric acid
C6H9ClN2O4S
MFCD00050933
20542670
C1=CC(=CC(=C1)Cl)NN.OS(=O)(=O)O

Specifications

Undesignated
41713-37-5
MFCD00050933
C1=CC(=CC(=C1)Cl)NN.OS(=O)(=O)O
240.658
383.24
≥98.0% (T)
5g
C6H9ClN2O4S
ZQMWFTLJZQAIHV-UHFFFAOYSA-N
(3-chlorophenyl)hydrazine;sulfuric acid
20542670
Crystal-Powder at 20°C
3-Chlorophenylhydrazine Sulfate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0471

TSCA : No