missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorophenylhydrazine hydrochloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 167361000

 View more versions of this product

Catalog No. AC167361000


 

Specifications Description & Specifications

Specifications

3-Chlorophenylhydrazine hydrochloride
100g
2312-23-4
100.0
Glass bottle
≥96.0%
C6H7ClN2·HCl
179.05
15, 424
White to pink or light beige
97%
96.0
Authentic
Powder
97%
ClC6H4NHNH2·HCl
MFCD00012935

Chemical Identifiers

2312-23-4
100.0
C6H7ClN2·HCl
179.05
3-Chlorophenylhydrazine hydrochloride, 97%
96.0
97%
ClC6H4NHNH2·HCl
MFCD00012935
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 219-005-6

TSCA : TSCA