missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorophenylhydrazine hydrochloride, 97%, ACROS Organics™

$331.68 - $331.68

Chemical Identifiers

CAS 2312-23-4
Molecular Formula C6H8Cl2N2
Molecular Weight (g/mol) 179.044
MDL Number MFCD00012935
InChI Key CRRIAWUJYMLJOE-UHFFFAOYSA-N
Synonym 3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride
PubChem CID 75331
IUPAC Name (3-chlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)NN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC167361000 SDS View Documents Acros Organics
167361000
100g Glass bottle Each for $331.68
AC167360250 SDS View Documents Acros Organics
167360250
25g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

2312-23-4
179.044
CRRIAWUJYMLJOE-UHFFFAOYSA-N
75331
C1=CC(=CC(=C1)Cl)NN.Cl
C6H8Cl2N2
MFCD00012935
3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride
(3-chlorophenyl)hydrazine;hydrochloride

Specifications

Authentic
Beige or Pink to White
2312-23-4
100.0
C6H8Cl2N2
MFCD00012935
3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride
C1=CC(=CC(=C1)Cl)NN.Cl
179.044
179.05
≥96.0%
Glass bottle
100g
96.0
97%
ClC6H4NHNH2·HCl
15, 424
CRRIAWUJYMLJOE-UHFFFAOYSA-N
(3-chlorophenyl)hydrazine;hydrochloride
75331
Powder
3-Chlorophenylhydrazine hydrochloride, 97%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 219-005-6

TSCA : TSCA