missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 167360250

Catalog No. AC167360250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Chlorophenylhydrazine hydrochloride
2312-23-4
100.0
96% min. (Argentometry)
C6H7ClN2·HCl
MFCD00012935
15, 424
Solubility in water: soluble. Other solubilities: soluble in methanol
C1=CC(=CC(=C1)Cl)NN.Cl
179.05
179.05
97%
96.0
Authentic
Glass bottle
ClC6H4NHNH2·HCl
25g
3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride
CRRIAWUJYMLJOE-UHFFFAOYSA-N
(3-chlorophenyl)hydrazine;hydrochloride
75331
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 219-005-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only