missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorophenylboronic acid, 97%, May contain varying amounts of anhydride, Maybridge

Chemical Identifiers

CAS 63503-60-6
Molecular Formula C6H6BClO2
Molecular Weight (g/mol) 156.372
MDL Number MFCD00161354
InChI Key SDEAGACSNFSZCU-UHFFFAOYSA-N
Synonym 3-chloro benzene boronic acid
PubChem CID 2734323
IUPAC Name (3-chlorophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)Cl)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC35900DA SDS View Documents Fisher Scientific Company
AC35900DA
1g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

63503-60-6
156.372
SDEAGACSNFSZCU-UHFFFAOYSA-N
2734323
B(C1=CC(=CC=C1)Cl)(O)O
C6H6BClO2
MFCD00161354
3-chloro benzene boronic acid
(3-chlorophenyl)boronic acid

Specifications

Authentic
Green-Yellow to White
63503-60-6
100.0
C6H6BClO2
3-chloro benzene boronic acid
B(C1=CC(=CC=C1)Cl)(O)O
156.372
156.38
≥96.0%
Amber Glass Bottle
1g
96.0
97%
MFCD00161354
SDEAGACSNFSZCU-UHFFFAOYSA-N
(3-chlorophenyl)boronic acid
2734323
Crystalline Powder or Needles
3-Chlorophenylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.