Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorophenylacetonitrile, 99%

Chemical Identifiers

Chemical Name or Material 3-Chlorophenylacetonitrile
CAS 1529-41-5
Molecular Formula C8H6ClN
Formula Weight 151.6
MDL Number MFCD00001908
UN Number UN3276
Beilstein 2042965
Synonym 3-Chlorobenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1567322

  SDS
alfa aesar™
A1567322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.6
CAS 1529-41-5
UN Number UN3276
Boiling Point 134° to 136°C (10mmHg)
Quantity 100g
Molecular Formula C8H6ClN
MDL Number MFCD00001908
Odor Pungent
Synonym 3-Chlorobenzyl cyanide
Melting Point 11° to 13°C
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Beilstein 2042965
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S
RTECS Number AL8240000
Chemical Name or Material 3-Chlorophenylacetonitrile
Assay 99%