missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorophenylacetonitrile, 99%

Manufacturer:  Alfa Aesar™ A1567322

 View more versions of this product

Catalog No. AAA1567322



Specifications

Specifications

3-Chlorophenylacetonitrile
Pungent
11°C to 13°C
1529-41-5
MFCD00001908
2042965
GTIKLPYCSAMPNG-UHFFFAOYSA-N
2-(3-chlorophenyl)acetonitrile
73722
99%
>110°C (230°F)
134°C to 136°C (10mmHg)
100g
C8H6ClN
UN3276
3-chlorobenzyl cyanide, 3-chlorophenyl acetonitrile, 2-3-chlorophenyl acetonitrile, 3-chlorophenylacetonitrile, 3-chlorobenzeneacetonitrile, m-chlorobenzyl cyanide, acetonitrile, m-chlorophenyl, benzeneacetonitrile, 3-chloro, m-chlorophenyl acetonitrile, 2-3-chlorophenyl ethanenitrile
C1=CC(=CC(=C1)Cl)CC#N
151.593
151.6

Chemical Identifiers

1529-41-5
151.593
GTIKLPYCSAMPNG-UHFFFAOYSA-N
73722
C1=CC(=CC(=C1)Cl)CC#N
C8H6ClN
MFCD00001908
3-chlorobenzyl cyanide, 3-chlorophenyl acetonitrile, 2-3-chlorophenyl acetonitrile, 3-chlorophenylacetonitrile, 3-chlorobenzeneacetonitrile, m-chlorobenzyl cyanide, acetonitrile, m-chlorophenyl, benzeneacetonitrile, 3-chloro, m-chlorophenyl acetonitrile, 2-3-chlorophenyl ethanenitrile
2-(3-chlorophenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S

RTECSNumber : AL8240000

Documents