Please login to your online account to display your discounted pricing

3-Chlorophenylacetone, 94%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35069-0050

 View more versions of this product

Catalog No. AC350690050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥92% (GC)
Physical Form Liquid
Formula Weight 168.62
Quantity 5g
Color Clear yellow
Chemical Name or Material 3-Chlorophenylacetone
Refractive Index 1.5320 to 1.5360 (20°C, 589nm)
Assay Percent Range 94%
Molecular Formula C9H9ClO
CAS 14123-60-5
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Chlorophenylacetone
Assay ≥92% (GC)
Assay Percent Range 94%
Molecular Formula C9H9ClO
Formula Weight 168.62