missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chlorophenylacetic acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC109840500

 View more versions of this product

Catalog No. AC109840500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥99.0%
170.6
White
09, II, 306
99.0
1878-65-5
MFCD00004332
Authentic
ClC6H4CH2CO2H
Shiny Flakes and Chunks
50g
77° to 80°C
100.0
3-Chlorophenylacetic acid
C8H7ClO2
Glass bottle
99+%

Chemical Identifiers

1878-65-5
100.0
C8H7ClO2
170.6
3-Chlorophenylacetic acid
99.0
99+%
ClC6H4CH2CO2H
MFCD00004332
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes severe skin burns and eye damage. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning