missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorophenyl Isocyanate, TCI America™

$56.53 - $632.52

Chemical Identifiers

CAS 2909-38-8
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.565
MDL Number MFCD00002016
InChI Key HHIRBXHEYVDUAM-UHFFFAOYSA-N
Synonym 3-chlorophenyl isocyanate, m-chlorophenyl isocyanate, benzene, 1-chloro-3-isocyanato, 3-chlorophenylisocyanate, 3-chloroisocyanatobenzene, isocyanic acid, m-chlorophenyl ester, m-chlorfenylisokyanat, isocyanic acid 3-chlorophenyl ester, m-chlorfenylisokyanat czech, 3-chlorobenzenisocyanate
PubChem CID 17968
IUPAC Name 1-chloro-3-isocyanatobenzene
SMILES C1=CC(=CC(=C1)Cl)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0119500G View Documents TCI America
I0119500G
500g Each for $632.52
 
I011925G View Documents TCI America
I011925G
25g Each for $56.53
 
Specifications

Chemical Identifiers

2909-38-8
153.565
HHIRBXHEYVDUAM-UHFFFAOYSA-N
17968
C1=CC(=CC(=C1)Cl)N=C=O
C7H4ClNO
MFCD00002016
3-chlorophenyl isocyanate, m-chlorophenyl isocyanate, benzene, 1-chloro-3-isocyanato, 3-chlorophenylisocyanate, 3-chloroisocyanatobenzene, isocyanic acid, m-chlorophenyl ester, m-chlorfenylisokyanat, isocyanic acid 3-chlorophenyl ester, m-chlorfenylisokyanat czech, 3-chlorobenzenisocyanate
1-chloro-3-isocyanatobenzene

Specifications

204°C
-4°C
2909-38-8
MFCD00002016
3-chlorophenyl isocyanate, m-chlorophenyl isocyanate, benzene, 1-chloro-3-isocyanato, 3-chlorophenylisocyanate, 3-chloroisocyanatobenzene, isocyanic acid, m-chlorophenyl ester, m-chlorfenylisokyanat, isocyanic acid 3-chlorophenyl ester, m-chlorfenylisokyanat czech, 3-chlorobenzenisocyanate
C1=CC(=CC(=C1)Cl)N=C=O
153.565
153.57
≥98.0% (GC)
Yellow
500g
C7H4ClNO
2206
HHIRBXHEYVDUAM-UHFFFAOYSA-N
1-chloro-3-isocyanatobenzene
17968
Clear Liquid at 20°C
3-Chlorophenyl Isocyanate
Safety and Handling

Safety and Handling

RTECSNumber : NQ8560000

TSCA : Yes

Recommended Storage : Refrigerator