Please login to your online account to display your discounted pricing

3-chlorophenyl isocyanate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  404961000

 View more versions of this product

Catalog No. AC404961000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 153.57
Physical Form Liquid
Quantity 100g
Color Clear colorless to slightly yellow
CAS Max % 100.0
Molecular Formula C7H4ClNO
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Linear Formula NCOC6H4Cl
CAS 2909-38-8
CAS Min % 97.5
Assay Percent Range 98%
MDL Number MFCD00002016
Chemical Name or Material 3-Chlorophenyl isocyanate
Infrared Spectrum Authentic
Refractive Index 1.5576 to 1.5596 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Name Note 98%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Specifications

Chemical Name or Material 3-Chlorophenyl isocyanate
Name Note 98%
CAS 2909-38-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4ClNO
Linear Formula NCOC6H4Cl
Formula Weight 153.57
MDL Number MFCD00002016
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents