missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorophenoxyacetonitrile, 98%

Manufacturer:  Alfa Aesar™ A1573014

 View more versions of this product

Catalog No. AAA1573014



Specifications

Specifications

3-Chlorophenoxyacetonitrile
28°C to 30°C
43111-32-6
MFCD00017332
3-chlorophenoxyacetonitrile, 2-3-chlorophenoxy acetonitrile, acetonitrile, 3-chlorophenoxy, acmc-1aqe2, 3-chloro-phenoxy-acetonitrile, nhhnupmhgviqqc-uhfffaoysa, 2-3-chlorophenoxy ethanenitrile, 2-3-chlorophenoxy-acetonitrile
C1=CC(=CC(=C1)Cl)OCC#N
167.592
167.6
126°C to 128°C (1mmHg)
25g
C8H6ClNO
UN3439
NHHNUPMHGVIQQC-UHFFFAOYSA-N
2-(3-chlorophenoxy)acetonitrile
142655
98%

Chemical Identifiers

43111-32-6
167.592
NHHNUPMHGVIQQC-UHFFFAOYSA-N
142655
C1=CC(=CC(=C1)Cl)OCC#N
C8H6ClNO
MFCD00017332
3-chlorophenoxyacetonitrile, 2-3-chlorophenoxy acetonitrile, acetonitrile, 3-chlorophenoxy, acmc-1aqe2, 3-chloro-phenoxy-acetonitrile, nhhnupmhgviqqc-uhfffaoysa, 2-3-chlorophenoxy ethanenitrile, 2-3-chlorophenoxy-acetonitrile
2-(3-chlorophenoxy)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents