missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorophenoxyacetic acid, 98+%

$78.34 - $260.62

Chemical Identifiers

CAS 588-32-9
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00015850
InChI Key XSBUXVWJQVTYLC-UHFFFAOYSA-N
Synonym 3-chlorophenoxyacetic acid, 2-3-chlorophenoxy acetic acid, 3-chlorophenoxy acetic acid, m-chlorophenoxyacetic acid, acetic acid, 3-chlorophenoxy, 3-chloro-phenoxy-acetic acid, acetic acid, m-chlorophenoxy, unii-jxk525064m, acmc-209m7g, 3-chlorophenoxy acetic acid
PubChem CID 11497
IUPAC Name 2-(3-chlorophenoxy)acetic acid
SMILES C1=CC(=CC(=C1)Cl)OCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0093906 View Documents Alfa Aesar™
L0093906
5g Each for $78.34
 
AAL0093914 View Documents Alfa Aesar™
L0093914
25g Each for $260.62
 
Specifications

Chemical Identifiers

588-32-9
186.591
XSBUXVWJQVTYLC-UHFFFAOYSA-N
11497
C1=CC(=CC(=C1)Cl)OCC(=O)O
C8H7ClO3
MFCD00015850
3-chlorophenoxyacetic acid, 2-3-chlorophenoxy acetic acid, 3-chlorophenoxy acetic acid, m-chlorophenoxyacetic acid, acetic acid, 3-chlorophenoxy, 3-chloro-phenoxy-acetic acid, acetic acid, m-chlorophenoxy, unii-jxk525064m, acmc-209m7g, 3-chlorophenoxy acetic acid
2-(3-chlorophenoxy)acetic acid

Specifications

107°C to 109°C
588-32-9
MFCD00015850
3-chlorophenoxyacetic acid, 2-3-chlorophenoxy acetic acid, 3-chlorophenoxy acetic acid, m-chlorophenoxyacetic acid, acetic acid, 3-chlorophenoxy, 3-chloro-phenoxy-acetic acid, acetic acid, m-chlorophenoxy, unii-jxk525064m, acmc-209m7g, 3-chlorophenoxy acetic acid
C1=CC(=CC(=C1)Cl)OCC(=O)O
186.591
186.59
3-Chlorophenoxyacetic acid
5g
C8H7ClO3
1911185
XSBUXVWJQVTYLC-UHFFFAOYSA-N
2-(3-chlorophenoxy)acetic acid
11497
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger