missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Chlorophenoxy)acetyl chloride, 95%

$123.51 - $493.47

Chemical Identifiers

CAS 114476-84-5
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD02094018
InChI Key KPINZNVDHXIYAR-UHFFFAOYSA-N
Synonym 3-chlorophenoxyacetyl chloride, 3-chlorophenoxy acetyl chloride, 2-3-chlorophenoxy acetyl chloride, acetyl chloride, 3-chlorophenoxy, acmc-1brdk, acetyl chloride,2-3-chlorophenoxy
PubChem CID 7010339
IUPAC Name 2-(3-chlorophenoxy)acetyl chloride
SMILES C1=CC(=CC(=C1)Cl)OCC(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1842703 View Documents Alfa Aesar™
L1842703
1g Each for $123.51
 
AAL1842706 View Documents Alfa Aesar™
L1842706
5g Each for $493.47
 
Specifications

Chemical Identifiers

114476-84-5
205.034
KPINZNVDHXIYAR-UHFFFAOYSA-N
7010339
C1=CC(=CC(=C1)Cl)OCC(=O)Cl
C8H6Cl2O2
MFCD02094018
3-chlorophenoxyacetyl chloride, 3-chlorophenoxy acetyl chloride, 2-3-chlorophenoxy acetyl chloride, acetyl chloride, 3-chlorophenoxy, acmc-1brdk, acetyl chloride,2-3-chlorophenoxy
2-(3-chlorophenoxy)acetyl chloride

Specifications

>110°C (230°F)
1g
C8H6Cl2O2
UN3265
3-chlorophenoxyacetyl chloride, 3-chlorophenoxy acetyl chloride, 2-3-chlorophenoxy acetyl chloride, acetyl chloride, 3-chlorophenoxy, acmc-1brdk, acetyl chloride,2-3-chlorophenoxy
C1=CC(=CC(=C1)Cl)OCC(=O)Cl
205.034
205.04
(3-Chlorophenoxy)acetyl chloride
120°C to 122°C (2mmHg)
114476-84-5
MFCD02094018
1871611
KPINZNVDHXIYAR-UHFFFAOYSA-N
2-(3-chlorophenoxy)acetyl chloride
7010339
95%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.