Learn More
3-Chlorophenol, 99%, Thermo Scientific Chemicals
$36.92 - $231.47
Chemical Identifiers
CAS | 108-43-0 |
---|---|
Molecular Formula | C6H5ClO |
Molecular Weight (g/mol) | 128.56 |
MDL Number | MFCD00002256 |
InChI Key | HORNXRXVQWOLPJ-UHFFFAOYSA-N |
Synonym | m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641 |
PubChem CID | 7933 |
ChEBI | CHEBI:38855 |
IUPAC Name | 3-chlorophenol |
SMILES | OC1=CC=CC(Cl)=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC109810050
|
Thermo Scientific Chemicals
109810050 |
5 g | Glass bottle |
Each for $36.92
|
|
||||
AC109811000
|
Thermo Scientific Chemicals
109811000 |
100 g | Glass bottle |
Each for $231.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-43-0 | |
128.56 | |
HORNXRXVQWOLPJ-UHFFFAOYSA-N | |
7933 | |
3-chlorophenol |
C6H5ClO | |
MFCD00002256 | |
m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641 | |
CHEBI:38855 | |
OC1=CC=CC(Cl)=C1 |
Specifications
108-43-0 | |
100.0 | |
Brown to Yellow | |
214.0°C | |
Authentic | |
Glass bottle | |
1.5650 to 1.5670 | |
MFCD00002256 | |
06,185 | |
15,2157 | |
Solubility in water: 27.7g/l (20°C). Other solubilities: soluble in alcohol,ether,benzene and aq. alkali | |
OC1=CC=CC(Cl)=C1 | |
128.56 | |
11.55 Pas (25°C) | |
128.56 | |
Liquid After Melting |
98.5 | |
33.0°C to 35.0°C | |
1.2180g/mL | |
97°C | |
99% | |
C6H5ClO | |
ClC6H4OH | |
5 g | |
1.218 | |
m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641 | |
HORNXRXVQWOLPJ-UHFFFAOYSA-N | |
3-chlorophenol | |
7933 | |
CHEBI:38855 | |
99% | |
3-Chlorophenol, 99% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in
GHS Signal Word: Warning
EINECSNumber : 203-582-6
RTECSNumber : SK2450000
TSCA : TSCA
RUO – Research Use Only