Please login to your online account to display your discounted pricing

3-Chlorophenethylalcohol, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC185960010

 View more versions of this product

Catalog No. AC185960010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 156.61
Quantity 1g
Color Clear colorless to very slightly yellow
EINECS Number 225-964-1
TSCA TSCA
Density 1.181g/mL
CAS Max % 100.0
MDL Number MFCD00002892
CAS Min % 97.5
Chemical Name or Material 3-Chlorophenethylalcohol
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H9ClO
Linear Formula ClC6H4CH2CH2OH
CAS 5182-44-5
Refractive Index 1.5475 to 1.5495 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Chlorophenethylalcohol
Name Note 98%
CAS 5182-44-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9ClO
Linear Formula ClC6H4CH2CH2OH
Formula Weight 156.61
MDL Number MFCD00002892