missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorophenethylalcohol, 98%, ACROS Organics™

Chemical Identifiers

CAS 5182-44-5
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002892
InChI Key NDWAVJKRSASRPH-UHFFFAOYSA-N
Synonym 2-3-chlorophenyl ethanol, 3-chlorophenethyl alcohol, benzeneethanol, 3-chloro, 3-chlorophenethylalcohol, 2-3-chlorophenyl ethan-1-ol, m-chlorophenethyl alcohol, m-chlorophenethylic alcohol, phenethyl alcohol, m-chloro, 3-chlorobenzeneethanol, acmc-1cuee
PubChem CID 78856
IUPAC Name 2-(3-chlorophenyl)ethanol
SMILES C1=CC(=CC(=C1)Cl)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC185960250 SDS View Documents Acros Organics
185960250
25g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC185960050 SDS View Documents Acros Organics
185960050
5g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

5182-44-5
156.609
NDWAVJKRSASRPH-UHFFFAOYSA-N
78856
C1=CC(=CC(=C1)Cl)CCO
C8H9ClO
MFCD00002892
2-3-chlorophenyl ethanol, 3-chlorophenethyl alcohol, benzeneethanol, 3-chloro, 3-chlorophenethylalcohol, 2-3-chlorophenyl ethan-1-ol, m-chlorophenethyl alcohol, m-chlorophenethylic alcohol, phenethyl alcohol, m-chloro, 3-chlorobenzeneethanol, acmc-1cuee
2-(3-chlorophenyl)ethanol

Specifications

1.1810g/mL
Authentic
1.5475 to 1.5495
135.0°C to 137.0°C (13.0mmHg)
25g
97.5
98%
ClC6H4CH2CH2OH
2-3-chlorophenyl ethanol, 3-chlorophenethyl alcohol, benzeneethanol, 3-chloro, 3-chlorophenethylalcohol, 2-3-chlorophenyl ethan-1-ol, m-chlorophenethyl alcohol, m-chlorophenethylic alcohol, phenethyl alcohol, m-chloro, 3-chlorobenzeneethanol, acmc-1cuee
C1=CC(=CC(=C1)Cl)CCO
156.609
156.61
98%
>110°C
Glass bottle
1.181
Colorless to Yellow
5182-44-5
100.0
C8H9ClO
MFCD00002892
NDWAVJKRSASRPH-UHFFFAOYSA-N
2-(3-chlorophenyl)ethanol
78856
Liquid
3-Chlorophenethylalcohol, 98%
Safety and Handling

Safety and Handling

EINECSNumber : 225-964-1

TSCA : TSCA