missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5
Supplier: Thermo Scientific Chemicals 255791000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
3-Chloroperoxybenzoic Acid | |
937-14-4,535-80-8,7732-18-5 | |
75.0,30.0,30.0 | |
White | |
di-m-Chlorobenzoyl peroxide: < 0.1% Typical (HPLC) | |
C7H5ClO3 | |
MFCD00002127 | |
3106 | |
01,135; 02,68; 03,49; 04,85; 05,120; 06,110; 07,62; 08,97; 09,108; 10,92; 11,122; 12,118; 13,76; 15,86; 16,80; 17,76 | |
15,2154 | |
Solubility in water: insoluble. Other solubilities: soluble in koh, soluble in chloroform and acetone | |
OOC(=O)C1=CC=CC(Cl)=C1 | |
172.56 | |
CHEBI:52091 | |
70-75% |
70 - 75% | |
70.0,25.0,25.0 | |
92.0°C to 94.0°C | |
0.5600g/mL | |
Plastic Bottle | |
ClC6H4CO3H | |
100 g | |
09,IV,972 | |
0.56 | |
3-chloroperoxybenzoic acid, 3-chloroperbenzoic acid, mcpba, m-chloroperbenzoic acid, m-chloroperoxybenzoic acid, meta-chloroperoxybenzoic acid, benzenecarboperoxoic acid, 3-chloro, m-chlorobenzoyl hydroperoxide, meta-chloroperbenzoic acid, 3-chlorobenzoperoxoic acid | |
NHQDETIJWKXCTC-UHFFFAOYSA-N | |
3-chlorobenzenecarboperoxoic acid | |
70297 | |
172.57 | |
Moist Powder |
RUO – Research Use Only