Please login to your online account to display your discounted pricing

3-(chloromethyl)heptane, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Chloromethyl)heptane
CAS 123-04-6
CAS Min % 88.0
CAS Max % 100.0
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C8H17Cl
Linear Formula H3C(CH2)3CH(CH2Cl)CH2CH3
Formula Weight 148.68
MDL Number MFCD00018940
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC344980250

  SDS
thermo fisher scientific
344980250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥88%
Physical Form Liquid
Formula Weight 148.68
Quantity 25mL
Color Clear yellow
CAS Min % 88.0
CAS Max % 100.0
Packaging Glass bottle
Linear Formula H3C(CH2)3CH(CH2Cl)CH2CH3
CAS 123-04-6
Synonym 2-Ethylhexylchloride
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Chemical Name or Material 3-(Chloromethyl)heptane
Refractive Index 1.4320 to 1.4360 (20°C, 589nm)
Assay Percent Range 90%
Molecular Formula C8H17Cl
MDL Number MFCD00018940
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.