missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Chloromethyl)benzoic acid, 97%

$17.59 - $140.80

Chemical Identifiers

CAS 31719-77-4
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00191922
InChI Key PBSUMBYSVFTMNG-UHFFFAOYSA-N
Synonym 3-chloromethyl benzoic acid, alpha-chloro-m-toluic acid, 3-chloromethyl-benzoic acid, benzoic acid, 3-chloromethyl, pubchem7446, acmc-209hog, 3-carboxybenzyl chloride, 3-chloromethyl benzoicacid, ksc497k4l, chembl67150
PubChem CID 241752
IUPAC Name 3-(chloromethyl)benzoic acid
SMILES C1=CC(=CC(=C1)CCl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6085402 View Documents Alfa Aesar™
H6085402
0.5g Each for $17.59
 
AAH6085409 View Documents Alfa Aesar™
H6085409
10g Each for $140.80
 
Description

Description

Applications
3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

31719-77-4
170.592
PBSUMBYSVFTMNG-UHFFFAOYSA-N
241752
C1=CC(=CC(=C1)CCl)C(=O)O
C8H7ClO2
MFCD00191922
3-chloromethyl benzoic acid, alpha-chloro-m-toluic acid, 3-chloromethyl-benzoic acid, benzoic acid, 3-chloromethyl, pubchem7446, acmc-209hog, 3-carboxybenzyl chloride, 3-chloromethyl benzoicacid, ksc497k4l, chembl67150
3-(chloromethyl)benzoic acid

Specifications

136°C to 138°C
31719-77-4
MFCD00191922
3-chloromethyl benzoic acid, alpha-chloro-m-toluic acid, 3-chloromethyl-benzoic acid, benzoic acid, 3-chloromethyl, pubchem7446, acmc-209hog, 3-carboxybenzyl chloride, 3-chloromethyl benzoicacid, ksc497k4l, chembl67150
C1=CC(=CC(=C1)CCl)C(=O)O
170.592
170.59
3-(Chloromethyl)benzoic acid
0.5g
C8H7ClO2
UN3261
PBSUMBYSVFTMNG-UHFFFAOYSA-N
3-(chloromethyl)benzoic acid
241752
97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.