missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Chloromethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6085402

 View more versions of this product

Catalog No. AAH6085402



Description

Description

3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.
Specifications

Specifications

3-(Chloromethyl)benzoic acid
136°C to 138°C
MFCD00191922
UN3261
Soluble in methanol.
OC(=O)C1=CC=CC(CCl)=C1
170.59
170.59
31719-77-4
C8H7ClO2
0.5 g
3-chloromethyl benzoic acid, alpha-chloro-m-toluic acid, 3-chloromethyl-benzoic acid, benzoic acid, 3-chloromethyl, pubchem7446, acmc-209hog, 3-carboxybenzyl chloride, 3-chloromethyl benzoicacid, ksc497k4l, chembl67150
PBSUMBYSVFTMNG-UHFFFAOYSA-N
3-(chloromethyl)benzoic acid
241752
97%

Chemical Identifiers

31719-77-4
170.59
PBSUMBYSVFTMNG-UHFFFAOYSA-N
241752
OC(=O)C1=CC=CC(CCl)=C1
C8H7ClO2
MFCD00191922
3-chloromethyl benzoic acid, alpha-chloro-m-toluic acid, 3-chloromethyl-benzoic acid, benzoic acid, 3-chloromethyl, pubchem7446, acmc-209hog, 3-carboxybenzyl chloride, 3-chloromethyl benzoicacid, ksc497k4l, chembl67150
3-(chloromethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only