Please login to your online account to display your discounted pricing

3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(Chloromethyl)-5-phenyl-1, 2, 4-oxadiazole
CAS 1201-68-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7ClN2O
Formula Weight 194.62
MDL Number MFCD00084968
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW02030CB

  SDS
fisher scientific company
SEW02030CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.62
Quantity 250mg
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD00084968
Chemical Name or Material 3-(Chloromethyl)-5-phenyl-1, 2, 4-oxadiazole
Molecular Formula C9H7ClN2O
CAS 1201-68-9