Please login to your online account to display your discounted pricing

3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW02030CB

 View more versions of this product

Catalog No. SEW02030CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 194.62
Quantity 250mg
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD00084968
Chemical Name or Material 3-(Chloromethyl)-5-phenyl-1, 2, 4-oxadiazole
Molecular Formula C9H7ClN2O
CAS 1201-68-9

Specifications

Chemical Name or Material 3-(Chloromethyl)-5-phenyl-1, 2, 4-oxadiazole
CAS 1201-68-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7ClN2O
Formula Weight 194.62
MDL Number MFCD00084968
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents