Please login to your online account to display your discounted pricing

3-(Chloromethyl)-5-methylisoxazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(Chloromethyl)-5-methylisoxazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6ClNO
Formula Weight 131.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB01262CB

 
fisher scientific company
SPB01262CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 131.56
Quantity 250mg
Assay 97%
Chemical Name or Material 3-(Chloromethyl)-5-methylisoxazole
Assay Percent Range 97%
CAS 35166-37-1
Molecular Formula C5H6ClNO
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le