Please login to your online account to display your discounted pricing

3-(chloromethyl)-5-(4-methoxyphenyl)-1,2,4-oxadiazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(Chloromethyl)-5-(4-methoxyphenyl)-1, 2, 4-oxadiazole
CAS 73217-31-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9ClN2O2
Formula Weight 224.65
MDL Number MFCD00084967
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW02033EA

  SDS
fisher scientific company
SEW02033EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.65
Quantity 10g
Assay 97%
CAS Min % 97.0
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 3-(Chloromethyl)-5-(4-methoxyphenyl)-1, 2, 4-oxadiazole
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Assay Percent Range 97%
MDL Number MFCD00084967
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C10H9ClN2O2
CAS 73217-31-9
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.