Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloromethyl-5-(3-methoxyphenyl)-1,2,4-oxadiazole, 99%

$99.14 - $340.53

Chemical Identifiers

Chemical Name or Material 3-Chloromethyl-5-(3-methoxyphenyl)-1,2,4-oxadiazole
CAS 175205-62-6
Molecular Formula C10H9ClN2O2
Formula Weight 224.65
MDL Number MFCD00084966
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5024303

  SDS
alfa aesar™
H5024303
1g Each for $340.53

AAH50243MD

  SDS
alfa aesar™
H50243MD
250mg Each for $99.14
Description & Specifications

Specifications

Formula Weight 224.65
Molecular Formula C10H9ClN2O2
Melting Point 58° to 59°C
Chemical Name or Material 3-Chloromethyl-5-(3-methoxyphenyl)-1,2,4-oxadiazole
CAS 175205-62-6
MDL Number MFCD00084966
Quantity 1g
Assay 99%