Please login to your online account to display your discounted pricing

3-(Chloromethyl)-5-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW06262CB

 View more versions of this product

Catalog No. SEW06262CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.65
Quantity 250mg
Assay 95%
CAS Min % 95.0
Chemical Name or Material 3-(Chloromethyl)-5-(2-thienyl)-1, 2, 4-oxadiazole
Packaging Amber Glass Bottle
Molecular Formula C7H5ClN2OS
CAS 306936-06-1
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD01571188
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-(Chloromethyl)-5-(2-thienyl)-1, 2, 4-oxadiazole
CAS 306936-06-1
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5ClN2OS
Formula Weight 200.65
MDL Number MFCD01571188
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents