missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Chloromethyl)-1-methyl-5-phenyl-1H-pyrazole, ≥97%, Maybridge

Chemical Identifiers

CAS 869901-13-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H11ClN2
Formula Weight 206.67
MDL Number MFCD08271938
Chemical Name or Material 3-(Chloromethyl)-1-methyl-5-phenyl-1H-pyrazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC46524CB SDS Fisher Scientific Company
CC46524CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

869901-13-3
100.0
C11H11ClN2
MFCD08271938
97%
96.5
97%
206.67
3-(Chloromethyl)-1-methyl-5-phenyl-1H-pyrazole

Specifications

869901-13-3
100.0
97%
C11H11ClN2
MFCD08271938
97%
96.5
Amber Glass Bottle
250mg
206.67
3-(Chloromethyl)-1-methyl-5-phenyl-1H-pyrazole

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger