Please login to your online account to display your discounted pricing

3-(Chloromethyl)-1-methyl-1H-pyrazole, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC48324DA

 View more versions of this product

Catalog No. CC48324DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 130.58
Quantity 1g
Assay 97%
CAS Max % 100.0
Molecular Formula C5H7ClN2
Assay Percent Range 97%
Chemical Name or Material 3-(Chloromethyl)-1-methyl-1H-pyrazole
MDL Number MFCD08690275
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.
CAS Min % 96.5
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement
If medical advice is needed, have product container or label at hand.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
CAS 84547-64-8

Specifications

Chemical Name or Material 3-(Chloromethyl)-1-methyl-1H-pyrazole
CAS 84547-64-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H7ClN2
Formula Weight 130.58
MDL Number MFCD08690275
Accessories

Danger

GHS P Statement
If medical advice is needed, have product container or label at hand.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.

SDS & Safety Documents