missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Chloromethyl)-1-methyl-1H-pyrazole, 97%, Maybridge

Chemical Identifiers

CAS 84547-64-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H7ClN2
Formula Weight 130.58
MDL Number MFCD08690275
Chemical Name or Material 3-(Chloromethyl)-1-methyl-1H-pyrazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC48324CB SDS Fisher Scientific Company
CC48324CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC48324DA SDS Fisher Scientific Company
CC48324DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

84547-64-8
100.0
C5H7ClN2
MFCD08690275
97%
96.5
97%
130.58
3-(Chloromethyl)-1-methyl-1H-pyrazole

Specifications

130.58
100.0
84547-64-8
Amber Glass Bottle
MFCD08690275
97%
250mg
C5H7ClN2
96.5
97%
3-(Chloromethyl)-1-methyl-1H-pyrazole

Safety and Handling

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement
If medical advice is needed, have product container or label at hand.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger