missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Chloromethyl)-1-methyl-1H-indazole, ≥97%, Maybridge

$139.86 - $139.86

Chemical Identifiers

CAS 1578-97-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H9ClN2
Formula Weight 180.64
MDL Number MFCD08060497
Chemical Name or Material 3-(Chloromethyl)-1-methyl-1H-indazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC06324CB SDS Fisher Scientific Company
CC06324CB
Amber Glass Bottle 250mg Each for $139.86
CC06324DA SDS Fisher Scientific Company
CC06324DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1578-97-8
100.0
C9H9ClN2
MFCD08060497
97%
96.5
97%
180.64
3-(Chloromethyl)-1-methyl-1H-indazole

Specifications

1578-97-8
100.0
97%
C9H9ClN2
MFCD08060497
97%
96.5
Amber Glass Bottle
250mg
180.64
3-(Chloromethyl)-1-methyl-1H-indazole

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger