Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloromandelic acid, 97%

$76.39 - $994.46

Chemical Identifiers

Chemical Name or Material 3-Chloromandelic acid
CAS 16273-37-3
Molecular Formula C8H7ClO3
Formula Weight 186.6
MDL Number MFCD00829409
Synonym 3-Chloro-alpha-hydroxyphenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2493706

  SDS
alfa aesar™
B2493706
5g Each for $76.39

AAB2493714

  SDS
alfa aesar™
B2493714
25g Each for $262.18

AAB2493722

  SDS
alfa aesar™
B2493722
100g Each for $994.46
Description & Specifications

Specifications

Formula Weight 186.6
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 5g
MDL Number MFCD00829409
Molecular Formula C8H7ClO3
Synonym 3-Chloro-alpha-hydroxyphenylacetic acid
CAS 16273-37-3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Chloromandelic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.