Please login to your online account to display your discounted pricing

3-chloroisonicotinic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC433750010

 View more versions of this product

Catalog No. AC433750010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (HPLC)
Formula Weight 157.56
Physical Form Solution
Quantity 1g
Color Brown to dark green to black
Melting Point 227° to 231°C
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C6H4ClNO2
Synonym 3-Chloropyridine-4-carboxylic acid
Chemical Name or Material 3-Chloroisonicotinic acid
Assay Percent Range 97%
MDL Number MFCD03094685
CAS 88912-27-0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning

Specifications

Chemical Name or Material 3-Chloroisonicotinic acid
CAS 88912-27-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C6H4ClNO2
Formula Weight 157.56
MDL Number MFCD03094685
Synonym 3-Chloropyridine-4-carboxylic acid
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents