missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chloroisonicotinic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 88912-27-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H4ClNO2
Formula Weight 157.56
MDL Number MFCD03094685
Synonym 3-Chloropyridine-4-carboxylic acid
Chemical Name or Material 3-Chloroisonicotinic acid
Assay ≥96.0% (HPLC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC433750010 SDS Thermo Fisher Scientific
AC433750010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC433750050 SDS Thermo Fisher Scientific
AC433750050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

88912-27-0
100.0
C6H4ClNO2
MFCD03094685
3-Chloroisonicotinic acid
96.0
97%
157.56
3-Chloropyridine-4-carboxylic acid
≥96.0% (HPLC)

Specifications

Brown to dark green to black
1g
88912-27-0
100.0
Solution
C6H4ClNO2
MFCD03094685
3-Chloroisonicotinic acid
227° to 231°C
Authentic
96.0
Glass bottle
97%
157.56
3-Chloropyridine-4-carboxylic acid
≥96.0% (HPLC)

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning