Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloroheptafluoro-2-butene, 96%

$59.38 - $59.38

Chemical Identifiers

Chemical Name or Material 3-Chloroheptafluoro-2-butene
CAS 434-41-3
Molecular Formula CF3CCl=CFCF3
Formula Weight 216.49
MDL Number MFCD00039261
Beilstein 1783461
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1239706

  SDS
alfa aesar™
L1239706
5g Each for $59.38
Description & Specifications

Specifications

Formula Weight 216.49
Chemical Name or Material 3-Chloroheptafluoro-2-butene
Assay 96%
MDL Number MFCD00039261
CAS 434-41-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula CF3CCl=CFCF3
Beilstein 1783461
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Boiling Point 31° to 32°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C