missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorobenzyl Chloride, 98+%, ACROS Organics™

$170.80 - $170.80

Chemical Identifiers

CAS 620-20-2
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C7H6Cl2
Linear Formula ClC6H4CH2Cl
Formula Weight 161.03
MDL Number MFCD00000905
Chemical Name or Material 3-Chlorobenzyl chloride
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC151701000 SDS Thermo Fisher Scientific
151701000
Glass bottle 100mL Each for $170.80
Specifications Description & Specifications

Chemical Identifiers

620-20-2
100.0
C7H6Cl2
161.03
3-Chlorobenzyl chloride
98.0
98+%
ClC6H4CH2Cl
MFCD00000905
≥98.0%

Specifications

Clear colorless to light yellow
620-20-2
100.0
Glass bottle
Liquid
C7H6Cl2
161.03
05, II, 231
≥98.0%
100mL
98.0
Authentic
1.5540 to 1.5560 (20°C, 589nm)
98+%
ClC6H4CH2Cl
MFCD00000905
3-Chlorobenzyl chloride